TipStation Pipette Tip Systems, Sorenson BioScience
Supplier: Sorenson Bioscience
27760T 27770T 27790 27790T
87000-762CS 526.09 CAD
87000-762 87000-764 87000-770 87000-772
TipStation Pipette Tip Systems, Sorenson BioScience
Pipette Tips
Stackable racks of tips are ideal for use with 8- and 12-channel pipettors.
  • Racks, lids, and base are manufactured from recycled plastic
Racks snap together and will not lift during pipetting. Recessed wells keep the tips stable for mounting on pipettors. The stacked design saves bench space and features a wide base to prevent the stack from tipping.
Ordering information: Each rack contains 192 tips in two 8×12 trays. The stack includes five racks of tips, one lid, and one base.