pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
BDH01880-55E
BDH50000-20F
BDH50120-20F
BDH5016-20ML
BDH50280-20F
BDH50360-20F
BDH50420-20F
BDH50440-20PK
BDH50680-20F
BDH50700-20PK
BDH50900-20F
BDH50960-20F
BDH0190-20L
BDH01920-55E
BDH0194-20L
BDH01960-55E
BDH0198-20L
BDH5002-500ML
BDH50040-20F
BDH5006-500ML
BDH5008-4L
BDH5010-500ML
BDH5014-500ML
BDH5018-500ML
BDH5020-4L
BDH5022-4L
BDH5024-500ML
BDH50260-4A
BDH50300-4A
BDH50320-500A
BDH5034-500ML
BDH5038-500ML
BDH5040-500ML
BDH5046-500ML
BDH5048-4L
BDH5050-4L
BDH5052-500ML
BDH50540-4A
BDH50560-20F
BDH50580-500A
BDH5060-500ML
BDH5062-500ML
BDH50640-4A
BDH5066-500ML
BDH5072-500ML
BDH5074-4L
BDH5076-4L
BDH5078-500ML
BDH50800-4A
BDH50820-20F
BDH5084-500ML
BDH5086-500ML
BDH5088-4L
BDH5092-500ML
BDH50940-4A
BDH50980-4A
BDH5100-PK
BDH0188-55GLEA
1966.89
CAD
BDH0188-55GL
BDH5000-20L
BDH5012-20L
BDH5016-20ML
BDH5028-20L
BDH5036-20L
BDH5042-20L
BDH5044-20ML
BDH5068-20L
BDH5070-20ML
BDH5090-20L
BDH5096-20L
BDH0190-20L
BDH0192-55GL
BDH0194-20L
BDH0196-55GL
BDH0198-20L
BDH5002-500ML
BDH5004-20L
BDH5006-500ML
BDH5008-4L
BDH5010-500ML
BDH5014-500ML
BDH5018-500ML
BDH5020-4L
BDH5022-4L
BDH5024-500ML
BDH5026-4L
BDH5030-4L
BDH5032-500ML
BDH5034-500ML
BDH5038-500ML
BDH5040-500ML
BDH5046-500ML
BDH5048-4L
BDH5050-4L
BDH5052-500ML
BDH5054-4L
BDH5056-20L
BDH5058-500ML
BDH5060-500ML
BDH5062-500ML
BDH5064-4L
BDH5066-500ML
BDH5072-500ML
BDH5074-4L
BDH5076-4L
BDH5078-500ML
BDH5080-4L
BDH5082-20L
BDH5084-500ML
BDH5086-500ML
BDH5088-4L
BDH5092-500ML
BDH5094-4L
BDH5098-4L
BDH5100-PK
pH Reference Standard Buffers, VWR Chemicals BDH®
Buffers
pH Calibration Buffers
BDH pH buffers are available as either colorless or colored solutions.
- Traceable to NIST Standards reference materials
- Calibrated in an ISO 17025 certified laboratory using certified Standards
- Store at room temperature
Ordering information: Buffer set BDH5100-PK comes with two 500 mL bottles each of the pH 4.00 red buffer, pH 7.00 yellow buffer, and pH 10.00 blue buffer.
SPECIFICATIONS
Environmentally Preferable |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...