Pure™ 1000G Pipette Tips, Molecular BioProducts
Supplier: Molecular BioProducts
Pure™
3590 3591 3592
53225-714CS 1904.98 CAD
53225-714 53225-716 53225-718
Pure™ 1000G Pipette Tips, Molecular BioProducts
Pipette Tips
The Pure 1000G tips are a larger volume version of the Pure 200G tips (53225-682).
  • Sterile tips are certified RNase-, DNase-, DNA-, and pyrogen-free
  • Non-sterile tips are autoclavable
  • Larger orifice design
Ideal for fragile cells, genomic DNA, and viscous samples. Compatible with Pipetman*, Eppendorf*, and others.
Ordering information: Tips 53225-718 and -716 are packaged 96 per tray, 10 trays per pack. Tips 53225-714 are bulk packaged.