Pure™ 200G Pipette Tips, Molecular BioProducts
Supplier: Molecular BioProducts
Pure™
3531
53225-682CS 1632.58 CAD
53225-682
Pure™ 200G Pipette Tips, Molecular BioProducts
Pipette Tips
The Pure 200G tips feature a large orifice designed for pipetting fragile cells, viscous samples, and genomic DNA.
  • Sterile tips are certified RNase-, DNase-, DNA-, and pyrogen-free
  • Nonsterile tips are autoclavable
Tips have a volume range of 40 to 200 µl for larger samples. Compatible with Pipetman*, Nichiryo/Benchmate*, Eppendorf*, and others.
Ordering information: Tips 53225-682 and -680 are packaged 96 per tray, 10 trays per pack. Tips 53225-678 are bulk packaged.