Pure™ 200 Pipette Tips, Molecular BioProducts
Supplier: Molecular BioProducts
Pure™
3552 3550 3551
53225-690CS 1740.27 CAD
53225-690 53225-692 53225-694
Pure™ 200 Pipette Tips, Molecular BioProducts
Pipette Tips
These tips feature the MicroPoint* design and a 10 µl reference mark for accurate sample handling and precise delivery.
  • Sterile tips are certified RNase-, DNase-, DNA-, and pyrogen-free
  • Nonsterile tips are autoclavable
Ideal for a wide variety of applications. Tips fit Pipetman*, Eppendorf*, Finnpipette*, Nichiryo*, Rainin*, and Transferpette* pipettors.
Ordering information: Tips 53225-694 and -692 are packaged 96 per tray, 10 trays per pack. Tips 53225-690 are bulk packaged.