Pure™ Ultra-Micro Pipette Tips, Molecular BioProducts
Supplier: Thermo Scientific
3520 3522
53225-671CS 1866.73 CAD
53225-671 53225-674 53225-676
Pure™ Ultra-Micro Pipette Tips, Molecular BioProducts
Pipette Tips
These tips are ideal for use with low volume samples.
  • Sterile tips are certified RNase-, DNase-, DNA-, and pyrogen-free
  • Non-sterile tips are autoclavable
The Ultra-Micro style allows for increased accuracy and repeatablility at low volumes. The longer tip length easily reaches sample tube bottoms. For use with Eppendorf*, Nichiryo*, and Transferpette* pipettors.
Ordering information: Tips 53225-674 are packaged 96 per tray, 10 trays per pack. Tips 53225-671 are bulk packaged.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO14001 (Certificate Registration No. EMS 752603)