Variable Transformers, STACO™
Supplier: Staco Energy Products
3PN1010B 3PN1010BV 3PN1510B 3PN2210B
62546-048EA 2287.21 CAD
62546-048 62546-053 62546-054 62546-056
Variable Transformers, STACO™
Transformers
Plug-in bench-type variable transformers provide linear, efficient non-distorted control and can be used wherever variable AC voltage is required
Units feature a ventilated steel case, a 1.8m (6') three-conductor grounded line cord, fused NEMA rated receptacle, and an illuminated on/off rocker switch. Graduated dial controls output voltage from 0 to 117% of the voltage setting. Metered unit 62546-053 comes with a pivot voltmeter with ±5% full scale accuracy, conveniently located atop the enclosure for easy readout.