Pierce™ Transfer Membranes, PVDF, Thermo Scientific
Supplier: Thermo Scientific
88518 88585 88520
CAPI88518EA 1098.05 CAD
CAPI88518 CAPI88585 CAPI88520
Pierce™ Transfer Membranes, PVDF, Thermo Scientific
Transfer Membranes
Hydrophobic transfer membranes made of high quality PVDF, which provides high binding capacity for proteins and nucleic acids.
  • Ideal for colorimetric and chemiluminescent detection
  • Resistant to discoloration and have better retention of absorbed protein
  • Lower background levels and increased sensitivity for fluorescent probing
Compatible with most organic solvents, acids and mild bases; doesn't tear or become brittle like nitrocellulose.
PVDF membranes offer a better retention of adsorbed proteins than other supports and are ideal for Western, Southern, Northern, and dot blots.