DURAN® protect+ Laboratory Bottles, Plastic Safety Coated, DIN 168-1 Thread, Graduated, Amber
Supplier: DWK Life Sciences (KIMBLE)
218061782 218061482 218062487 218063689 218064488 218065484 218066386 218067382
77575-028CS 988.94 CAD
77575-028 77575-026 77575-030 77575-032 77575-034 77575-036 77575-038 77575-040
DURAN® protect+ Laboratory Bottles, Plastic Safety Coated, DIN 168-1 Thread, Graduated, Amber
Bottles Glass Bottles
Multi-functional ambered glass laboratory bottles with UV protection and a protective safety coating.
  • Manufactured from ambered borosilicate glass 3.3 and polyamide
  • Standardised GL-thread compatible with matching cap system for particularly tight sealing and simple, clean pouring out
  • The protect+ safety coating provides scratch, leak*, and splinter* protection (*only applies to bottles 2000 ml and less)
  • UV protection USP <660> (Spectral Transmission) and EP (3.2.1) compliant; less than 10% spectral transmission between 290 and 450 nm
  • External only application of the amber colour and safety coating ensures that internal surfaces have standard DURAN® glass properties
  • Easy-to-read scale with graduation marks to estimate the content
  • 25 ml bottle has a specially shaped glass edge for improved pouring (additional plastic pouring ring is not required)
  • Safe storage and shipping of bulk, high-potency active pharmaceutical ingredients (HPAPIs) powders
  • Coating process does not use any volatile organic compounds VOCs or isocyanates
DURAN® GL ISO bottles have become the gold standard of multi-functional laboratory bottles since their introduction in 1972. The protect+ safety coated version of the bottles combines the best of both worlds: virtually inert USP/EP Type I borosilicate glass 3.3 on the inside and a protective safety plastic layer on the outside. The bottles are moulded from high-purity, clear glass or externally ambered clear glass that is then coated with a translucent polyamide PA 12 coating to reduce the hazards of broken glass and liquid splashes in the event of breakage.
A combination of high-performance materials and robust design provides a cost-effective usable life. The bottles feature an easy-to-read graduated volume scale. There is a retrace code printed on each bottle for manufacturing lot traceability. The amber DURAN® glass bottle versions ensure that light sensitive contents are protected from the effects of photodegradation. The protection against ultra-violet light meets international USP/EP standards. All the protect+ coated bottles are supplied with push-on protective covers to protect the inside of the bottles from contamination during distribution. Matching screw closures and pouring rings can be ordered separately.
Available in a wide range of nominal bottle capacities from 25 ml to 5 L, and neck sizes GL 25 to GLS 80® wide mouth. This bottle series is not autoclavable.
Delivery information: This laboratory bottle series is supplied as safety coated glass bottle only. Screw caps and pouring rings must be ordered separately.