ART® 300 Non-Filter Pipette Tips, Thermo Scientific
Supplier: Thermo Scientific ART
Pure™ SoftFit™
3772-B 3771 3772-05-B
89413-290CS 563.32 CAD
89413-290 34600-100 89413-288
ART® 300 Non-Filter Pipette Tips, Thermo Scientific
Pipette Tips
The design of these tips ensures a reliable fit across all channels for precise tip-to-tip aspiration.
  • Sterile tips are certified RNase-, DNase-, DNA-, and pyrogen-free
  • Nonsterile tips are autoclavable
Single motion loading minimizes hand fatigue. The reinforced, wide bottom tray provides extra stability and reliable tip attachment. Compatible with most single and multi-channel pipettors.
Ordering information: Tips 34600-100 and -102 are packaged 96 per tray, 10 trays per pack. Tips 89413-288 and is bulk packaged.