DURAN® pressure plus+ protect+ Laboratory Bottles, Plastic Safety Coated, DIN 168-1 GL 45 Thread, Graduated, Clear
Supplier: DWK Life Sciences (KIMBLE)
218155481 218152484 218153686 218154485
77575-048CS 1651.32 CAD
77575-048 77575-042 77575-044 77575-046
DURAN® pressure plus+ protect+ Laboratory Bottles, Plastic Safety Coated, DIN 168-1 GL 45 Thread, Graduated, Clear
Bottles Glass Bottles
Pressure and vacuum resistant glass laboratory bottles with plastic safety coating.
  • Manufactured from borosilicate glass 3.3, PA 12
  • Polyamide 12 safety coating provides scratch, leak, and splinter protection in the event of breakage due to impact
  • Plastic safety coating provides an enhanced safer grip
  • Special geometry still conforms to ISO 4796-1:2016 to maintain instrument compatibility
  • Optimum safety for working under pressure and suitable for use under a high vacuum
  • Fired-on, highly durable blue ceramic for visual differentiation from the standard
More Product Information