Nunc® 96-Well DeepWell Plates with Shared-Wall Technology, Thermo Scientific
Supplier: Thermo Fisher Scientific
278743 260251 260252
CA73520-474CS 1164.63 CAD
CA73520-474 CA73520-120 CA73520-122
Nunc® 96-Well DeepWell Plates with Shared-Wall Technology, Thermo Scientific
Microplates Deep Well Microplates
Nunc™ DeepWell plates with shared-wall technology are ideal for sample collection, storage, combinatorial chemistry, and library applications
They are resistant to most chemicals, solvents, and alcohols used in combinatorial chemistry. Shared-wall technology provides increased well volume, optimizing storage capacity and improved mixing. Plates feature cylindrical wells with round bottoms for optimal mixing and recovery. Alphanumeric grids are included for quick sample identification. All microplates are non-treated and supplied without lids.
Ordering information: Sterile plates are supplied individually wrapped. Nonsterile plates are supplied in packs of five, 12 packs per case.