Nunc® 96-Well Deep Well Plates, Thermo Scientific
Supplier: Thermo Fisher Scientific
278606 278605
CA44200-024CS 593.12 CAD
CA44200-024 CA44200-026
Nunc® 96-Well Deep Well Plates, Thermo Scientific
Microplates Deep Well Microplates
These clear, untreated plates are ideal for sample collection, library storage, combinational chemistry and HTS applications
Made of polystyrene resin. Designed for use with automatic and robotic handling instruments. Round bottom wells with a working volume of 0.9mL. Alphanumeric grid for quick and easy sample identification.