Axygen® MAXYMum Recovery™ Research-Grade Pipette Tips, Corning
Supplier: Corning
T-350-C-L T-300-L-STK TXL-10-L TXL-10-L-R TXL-10-L-R-S T-200-C-L T-200-C-L-R T-200-C-L-R-S T-200-C-L-STK TR-222-C-L TR-222-C-L-R TR-222-C-L-R-S TR-222-C-L-STK T-1000-C-L T-1000-C-L-R T-1000-C-L-R-S T-300-L T-300-L-R T-300-L-R-S T-400-L T-400-L-R T-400-L-R-S T-350-C-L-R-S T-350-C-L-R T-400-R-S
10011-011CS 1652.25 CAD
10011-011 10011-872 12000-208 12000-210 12000-212 22234-062 22234-064 22234-066 22234-068 22234-070 22234-072 22234-074 22234-076 22234-086 22234-088 22234-090 22234-092 22234-094 22234-096 22234-098 22234-100 22234-102 22234-128 22234-130 CA10011-347
Axygen® MAXYMum Recovery™ Research-Grade Pipette Tips, Corning
Pipette Tips
For applications requiring high accuracy and reproducibility.
  • Ideal for transferring viscous DNA, RNA, and protein solutions
  • Autoclavable under normal autoclaving guidelines
Tips are constructed using diamond-polished molds to eliminate the cavities and occlusions that cause sample retention. Made of a unique polymer that resists DNA, RNA, and protein adherence, tips will not denature DNA. Manufactured without any secondary substances such as fillers, silicone, or heavy metals, which can contaminate samples.
Presterilized tips are exposed to e-beamMore Product Information