MµltiFit™ Micro Volume Pipette Tips, Sorenson BioScience
Supplier: Sorenson Bioscience
13980T 23560T 23570T 23580T 30470T 37640 37640T 37650 37650T 37660 37820T 22210 22220-E 13980
14229-764CS 748.28 CAD
14229-764 14229-858 14229-860 14229-862 14229-878 14229-924 14229-926 14229-928 14229-930 14229-932 14229-938 53550-001 53550-004 53550-006
MµltiFit™ Micro Volume Pipette Tips, Sorenson BioScience
Pipette Tips
These natural-colored tips fit most popular brand research-grade pipettors.
They feature a narrow bore that reduces dead air space around extended air-displacement piston. Narrow bore also increases accuracy and reproducibility for critical ultra microvolume ranges.
Certifications: Certified RNase-, DNase-, human DNA-, PCR inhibitor-, and pyrogen-free.
Ordering information: Available in autoclavable polypropylene racks, bulk packed, or QuickRack reloads for the reloading racks.