Snapwell™ Inserts, Sterile, Corning
Supplier: Corning
3407 3801
29442-160CS 353.55 CAD
29442-160 29444-282
Snapwell™ Inserts, Sterile, Corning
Microplates Insert Plates and Inserts
A modified Transwell® permeable support containing a 12 mm Ø membrane supported by a detachable ring. Once cells are grown to confluence on the Snapwell™ insert, the ring can be placed in a vertical or horizontal diffusion chamber. Packaged in six well plates.
Ordering information: Snapwell™ inserts are supplied preloaded into 6-well plates.