SensoPlate™ 384-Well Glass-Bottom Plates, Greiner Bio-One
Supplier: Greiner Bio-One
781892 788892 781892P 781892-946
82051-350CS 992.07 CAD
82051-350 82051-488 82051-546 89093-110
SensoPlate™ 384-Well Glass-Bottom Plates, Greiner Bio-One
Microplates HTS Microplates
Microplates exhibit a bottom flatness better than 100 µm to offer high-quality performance for all applications where low autofluorescence and exceptional optical clarity are required.
  • Ideal for applications requiring low autofluorescence
  • Exceptional optical clarity
  • Sterile
  • Supplied with a lid
Ideal for high-resolution imaging, sensitive fluorescence, and confocal microscopy such as single molecule detection (SMD). Transmission measurements are capable in the wavelength range above 350nm. Plates consist of a 175 µm thick borosilicate glass-bottom plate bonded with a black polystyrene frame. Plate 82051-488 features a LoBase format. All plates are sterile, comply with SBS recommendations, and include a lid.
Ordering information: For 384-well SensoPlate™ Plus glass-bottom plates, see 82051-616 series.