Pierce™ Protein IgG Plus Orientation Kit, Thermo Scientific
Supplier: Thermo Scientific
Pierce™ Protein IgG Plus Orientation Kits uses a crosslinker to covalently attach antibody that is bound to Protein A (or Protein G) agarose resin, thereby creating a reusable affinity column for purification of antigens.
- Contaminant free antigen purification and less antibody leaching
- Covalent, leak resistant antibody immobilization
- Gentle binding and crosslinking conditions
- Less antibody leaching
- Crosslinked 6% beaded agarose with recombinant Protein A (or Protein G) thus making it high quality resin
- Oriented immobilization that is able to capture passing antigen in the mobile phase without steric hindrance
Delivery information: Kit includes Recombinant Protein A [or Protein G] columns, 2 mL (2 columns), Disuccinimidyl suberate [DSS] (2 x 13 mg), PBS pack (makes 500 mL), Blocking Buffer (6 mL), IgG Elution Buffer (15 mL), Antibody Binding/Wash Buffer (120 mL), and column accessories.