SensoPlate™ 24-Well Glass-Bottom Plate, Greiner Bio-One
Supplier: Greiner bio-one
SensoPlate™
662892
82050-898CS 456.9 CAD
82050-898
SensoPlate™ 24-Well Glass-Bottom Plate, Greiner Bio-One
Microplates HTS Microplates
Microplate exhibits a bottom flatness better than 100 µm to offer high-quality performance for all applications where low autofluorescence and exceptional optical clarity are required.
  • Ideal for applications requiring low autofluorescence
  • Exceptional optical clarity
  • Sterile
  • Chimney-well prevents cross-contamination
Ideal for high-resolution imaging, sensitive fluorescence, and confocal microscopy such as single molecule detection (SMD). Transmission measurements are capable in the wavelength range above 350nm. Plate consists of a 175 µm thick borosilicate glass-bottom plate bonded with a black polystyrene frame and features a chimney-well to help prevent cross-contamination. Plate is sterile, complies with SBS recommendations, and includes a lid.