Pierce™ Zeba™ Spin Desalting Plates, Thermo Scientific
Supplier: Thermo Scientific
89807 89808
CAPI89807EA 838.41 CAD
CAPI89807 CAPI89808
Pierce™ Zeba™ Spin Desalting Plates, Thermo Scientific
Microplates HTS Microplates
The Zeba™ spin desalting plates are polypropylene devices containing high-performance Zeba™ desalting resin, offering rapid protein desalting with high recovery of proteins >7000 daltons
  • High recovery from low-binding resin that maximizes protein recovery
  • One plate can process 96 small sample volumes (20 to 100 µL) in 5 minutes - no fraction screening or waiting for protein to emerge by gravity-flow
Zeba™ desalting devices provide trouble-free desalting and buffer exchange for sample volumes ranging from 2 µL to 4 mL. The 96-well spin plate format enables more high-throughput processing. Zeba™ 96-well spin desalting plates require no resin dispensing or hydration and provide the same high protein recovery as spin columns with at least 95% removal of salts and other small molecules.
Delivery information: Kit contains 2 (or 4) plates, each well contains ~550 µL resin slurry in 0.05% sodium azide and can process 20 - 100 µL samples; 2 wash plates; and 2 (or 4) collection plates.