Whatman™ Lids, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™ UniSeal™ UniSeal™
7704-1001 7704-1001
CA28317-488PK 396.75 CAD
CA28317-488 28317-488
Whatman™ Lids, Whatman products (Cytiva)
Lids Microplate Lids
Universal lids suitable for robotic handling. Can be used as dust covers and to prevent splashing or contamination when plates are being moved around the laboratory.