Proline® Plus Multichannel Mechanical Pipettors, Sartorius
Supplier: Sartorius
728120 728130 728140 728220 728230 728240 728120 728130 728140 728220 728230 728240
89082-290EA 1879.17 CAD
89082-290 89082-292 89082-294 89082-296 89082-298 89082-300 CA89082-290 CA89082-292 CA89082-294 CA89082-296 CA89082-298 CA89082-300
Proline® Plus Multichannel Mechanical Pipettors, Sartorius
Pipettes Micropipettes
Proline® Plus mechanical pipettors are designed to offer comfort and quality for everyday manual pipetting.
  • Ergonomic design with light pipetting forces, comfortable handle, and finger support for reduced risk of strain injuries
  • Minimized risk of contamination with Safe-Cone Filters and full autoclavability
  • Highly durable with strengthened structure for heavier use
Pipettors offer good UV resistance and excellent chemical resistance. The devices are suitable for both left- and right-handed users. Thermal insulation of internal components ensures that the warmth of the hand does not affect pipetting results during extended use. Pipettors are fully autoclavable. To facilitate tip selection, color coding on the pipettor's display corresponds to color coded tip trays.
They combine durable construction with ease and lightness of use, and are therefore appropriate for liquid-handling professionals and students alike.