Nunc® ShallowWell 384-Well Plates, Standard Height, Thermo Scientific
Supplier: Thermo Fisher Scientific
267460 267461 267462
CA55710-312CS 2568.83 CAD
CA55710-312 CA55710-314 CA55710-316
Nunc® ShallowWell 384-Well Plates, Standard Height, Thermo Scientific
Microplates HTS Microplates
Ideal for small volume applications, these polypropylene ANSI standard plates are automation compatible
Plates feature an untreated, low-binding, and chemical-resistant surface. Plates have square wells with round bottoms for optimal sample recovery and minimal wicking. Natural plates are ideal for storage or colorimetric assays, black plates are ideal for fluorescent assays, and white plates are suitable for luminescent assays.
Order Now
SPECIFICATIONS
Environmentally Preferable