Hygromycin B ≥80% (by HPLC), Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: (3aS,3'R,4S,4'S,5'R,6R,6'R,7S,7aS)-4-(((1R,2S,3R,5S,6R)-3-Amino-2,6-dihydroxy-5-(methylamino)cyclohexyl)oxy)-6'-((S)-1-amino-2-hydroxyethyl)-6-(hydroxymethyl)octahydro-3aH-spiro[[1,3]dioxolo[4,5-c]pyran-2,2'-pyran]-3',4',5',7-tetraol

400051-10MU 400051-1MU 400051-5MU 400051-100KU
CA80055-284EA 3288.56 CAD
CA80055-284 CA80055-286 CA80055-290 CA80603-022
Hygromycin B ≥80% (by HPLC), Sigma-Aldrich®
Hygromycin B

Caramel-colored liquid


  • Bioassay: 350–450 units/mg


Unit Definition: 106 units is equivalent to approximately 900mg.


Store at 4°C (39°F). Shipped as hazardous.


Warning: Highly toxic.


Unique aminoglycoside antibiotic that inhibits the growth of prokaryotic (bacteria) and eukaryotic microorganisms (yeasts) and mammalian cells. Inhibits protein synthesis at the translocation step on the 70S ribosome and causes misreading of the mRNA. Hph, a gene from Escherichia coli, encodes resistance to hygromycin B and can be isolated and cloned by recombinant DNA techniques. This hygromycin B-resistance gene is particularly useful for identification or selection of recombinant clones in a variety of cell types. Hygromycin B penetrates cells that have been permeabilized by virus infection, and can act as an effective antiviral agent.

Formula: C₂₀H₃₇N₃O₁₃
MW: 527.5 g/mol
MDL Number: MFCD06795479
CAS Number: 31282-04-9
Merck Index: 13,4878

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR