Top DNA Polymerase, Bioneer
Supplier: BIONEER, INC.
E-3100 E-3100-1 E-3100-2 E-3100-3 E-3101 E-3101-1 E-3101-2 E-3101-3
95027-542EA 196.49 CAD
95027-542 95027-544 95027-546 95027-548 95027-550 95027-552 95027-554 95027-556
Top DNA Polymerase, Bioneer
Nucleic Acid Reagents End-point PCR Enzymes and Kits
The versatile selection of top DNA polymerase offers improved base incorporation ability by inactivating 5' to 3' exonuclease activity
Polymerase is isolated from a recombinant E.coli strain containing the DNA polymerase gene from Thermus thermophilus. It exhibits highest activity at pH 9.0 and 72°C. Suitable for a variety of applications, such as standard PCR, primer extension, TA cloning, and gene sequencing.
Ordering information: One Bioneer unit is defined as the amount of enzyme required to incorporate 10nmol of dNTP into acid-insoluble material in 30 minutes at 72°C.