Nunc® 96-Well Optical Bottom Plates, Thermo Scientific
Supplier: Thermo Fisher Scientific
265301 265302
CA37000-550CS 560.28 CAD
CA37000-550 CA37000-544
Nunc® 96-Well Optical Bottom Plates, Thermo Scientific
Microplates HTS Microplates
These nonsterile polystyrene plates feature white or black upper structures with optically clear bottoms
The lids for optical bottom plates feature cut-off corners at right top and bottom.
The plates can be read from the top or bottom. The white plates are ideal for fluorescence or luminescence applications; the black plates are ideal for fluorescence applications. The polystyrene upper structure is fitted with a polystyrene film. The 96-well footprint makes the plates compatible with automated handling equipment. Plates are optimized for scintillation counting and feature alphanumericMore Product Information