Whatman™ UNIPLATE™ Collection and Analysis Microplates
Supplier: Cytiva
7701-1651 7701-3250 7701-5102 7701-5110 7701-5500 7701-5200 7701-1651 7701-3250 7701-5102 7701-5110 7701-5500 7701-5200
13503-152PK 490.06 CAD
13503-152 13503-170 13503-188 13503-190 13503-204 28317-472 CA13503-152 CA13503-170 CA13503-188 CA13503-190 CA13503-204 CA28317-472
Whatman™ UNIPLATE™ Collection and Analysis Microplates
Microplates HTS Microplates
The UNIPLATE™ series includes microplates with a range of well profiles, well volumes and well densities, made from diverse polymer materials. Most UNIPLATE™ microplates conform to the proposed SBS microplate standard and fit most microplate readers and automated plate handling devices.
They are suitable for a wide range of applications, including simple filtrate collection, when used in conjunction with UNIFILTER® microplates, as well as homogeneous assay techniques utilised in HTS.