Storage Microplates 384-well Polypropylene, Corning®
Supplier: Corning
Corning®
3658 3965 3657 3347 3656 3964 3342
29445-120CS 647.5 CAD
29445-120 29445-114 29445-136 29445-116 80081-116 15551-254 80081-126
Storage Microplates 384-well Polypropylene, Corning®
Microplates HTS Microplates
These 384-well polypropylene plates and blocks are built to the industry standard footprint
Robolid combines the sealing ability of a storage mat with the rigidity of a plastic lid and is designed for repeated application and removal by automation and to prevent short-term evaporation. Robolid also features silicone sealing plugs for organic solvent resistance and low extractables.
They are automation-friendly and solvent-resistant. They feature round-, conical-, or v-bottoms for optimal liquid handling and a rounded square-well geometry for reduced wicking. Both plates and blocks are certified DNase- and RNase-free.
Ordering information: The 384-well plates are packaged 25 per pack, except for 46610-064 which is packaged 10 per pack. The 384-well blocks are packaged 5 per pack.