HTS Transwell® 24-Well Permeable Supports System, Polystyrene, Sterile, Corning
Supplier: Corning
3396 3397 3398 3399 3379 3378
29445-098CS 380.66 CAD
29445-098 29445-100 29445-102 29445-104 46610-178 66025-540
HTS Transwell® 24-Well Permeable Supports System, Polystyrene, Sterile, Corning
Microplates HTS Microplates
HTS insert plates are arrays of individual cell culture inserts connected by a rigid, robotics-friendly holder. This single-unit design makes insert plates ideal for running automated, high throughput drug transport (Caco-2 cells) cell toxicity studies or cell migration and invasion studies.
  • Cell growth area is 0.33 cm²/well
  • Choice of either polyester (PET) membrane (0.4 µm pore size) or polycarbonate (PC) membrane (0.49 m and 3.0 µm pore sizes)
  • Treated for optimal cell attachment
  • Sterile
The HTS Transwell-24 permeable support has an array of 24 wellsMore Product Information