Protease Inhibitor Cocktail Set V, EDTA-Free, Calbiochem®
Supplier: MilliporeSigma
The cocktail consists of four protease inhibitors designed for the inhibition of serine, cysteine, but not metalloproteases.
Recommended for use with mammalian cell lysates and tissue extracts. When reconstituted with 1 ml H₂O, each vial contains a 100X stock solution. When diluted to 1X the cocktail contains the following components; AEBSF Hydrochloride (500 µM), Aprotinin Bovine Lung (150 nM), E-64 Protease Inhibitor (1 µM), and Leupeptin Hemisulfate (1 µM). Stock solutions are stable for up to six months at −20 °C (−4 °F). (EMD Biosciences 539137-10VL)
Ordering information: Each order is supplied as a set of 10×1 ml vials.