Ovation® E8 & E12, Electronic Multichannel Pipettors, VistaLab
Supplier: VistaLab
1060-0020 1060-0125 1060-0250 1060-0850 1160-0020 1160-0125 1160-0250 1160-1250
49000-548EA 3126.66 CAD
49000-548 49000-550 49000-552 49000-554 89002-376 89002-378 89002-380 89002-382
Ovation® E8 & E12, Electronic Multichannel Pipettors, VistaLab
Pipettes Micropipettes
These adjustable volume, air displacement, fully electronic motor-driven pipettors are intended to aspirate and dispense precise fluid volumes.
  • Contoured, natural shape gives the user maximum stability when pipetting, requiring minimal contact pressure
  • An adjustable hook provides custom comfort for left-handed and right-handed users
Pipettors feature LCD display and single keystroke pushbutton controls for easy access to liquid handling functions, calibration, settings, status, and remaining battery power. Acquire tips with only a gentle push; audibleMore Product Information