Socorex Acura® manual 855 Multichannel Pipettors, Variable Volume, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W810603 W810600 W810602 W810604 W810606 W810608 W810610 W810609
89044-616EA 3123.81 CAD
89044-616 83009-176 83009-178 83009-180 83009-182 83009-184 83009-186 89044-618
Socorex Acura® manual 855 Multichannel Pipettors, Variable Volume, DWK Life Sciences
Pipettes Micropipettes
These economical variable volume pipettors accurately transfer liquids to 96-well plates.
  • Soft-touch plunger reduces user fatigue
  • Autoclavable, while fully assembled, to 121 °C (250 °F)
Easy, swift-set calibration requires no additional tools beyond a balance and distilled water. Reliable volume adjustment with a turn of the push button, even with one hand. The JusTip™ adjustable tip ejector allows the use of most pipette tips. Fine click-stops and a freely rotating button cap prevent accidental volume changes. Hand hook permits the user to hold theMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable