Nunc® Immobilizer™ Amino Plates and Modules, Thermo Scientific
Supplier: Thermo Fisher Scientific
436006 436013 436023 436007 436008
CA73521-124CS 1435.6 CAD
CA73521-124 CA73521-126 CA73521-128 CA73521-254 CA73521-256
Nunc® Immobilizer™ Amino Plates and Modules, Thermo Scientific
Microplates Immunology Microplates and Strips
These plates are ideal for one-step covalent coupling of peptides, proteins, and aminated oligos with minimal denaturation
Clear plates guarantee uniform well-to-well binding capacity of CV ≤5%; black and white plates guarantee uniform well-to-well binding capacity of CV ≤7.5%.
Free amino or subhydryl groups on the biomolecule react spontaneously and are covalently tethered to the surface through an optimized spacer arm. Choose from polystyrene plates in standard 96-well format or polystyrene modules consisting of twelve 8-well strips. LockWell™ modules feature 8-well break apart strips that can be separated into individual wells and then locked into a frame at the same horizontal level for uniform washing and reading. Plates have a high signal-to-noise ratio, no surface leaching, reduced steric hindrance of bound molecules, and highly reproducible binding with low CVs. They require no blocking or activation steps and feature site directed binding. Plates are also stable for long periods at room temperature. Choose clear plates and strips for colorimetric assays, white plates for bioluminescence and chemiluminescence assays, and black plates for fluorescence assays.