VWR® Prefilled Histology Containers, 10% Neutral Buffered Formalin (NBF)
Supplier: Azer Scientific Inc.
PFNBF-20 16004-122 16004-124 16004-117 16004-121 16004-119
16004-112CS 579.69 CAD
16004-112 16004-122 16004-124 16004-117 16004-121 16004-119
VWR® Prefilled Histology Containers, 10% Neutral Buffered Formalin (NBF)
Sample Containers General Purpose Containers
Containers are half-filled with 10% neutral buffered formalin, for the collection and transport of tissue samples.
  • High clarity virgin polypropylene container construction
  • Polyethylene screw caps included
  • 20–120 mL containers are 95kPa compliant
Each container is made of high clarity virgin polypropylene and includes a polyethylene screw cap
Small-sized prefilled formalin containers (20 mL and 40 mL) feature a click-top closure. The first click assures users the seal is fully engaged. The second click ensures 95kPa compliance. This click-top design prevents leaking formalin containers and ensures safe specimen transport.
Containers sized 20 mL through 120 mL are packaged in transport packs with handles. This packaging design allows for easy distribution to other labs and doctors’ offices. The inner packaging is made of chipboard material, and is safe for operating room use.
Caps feature dual thread closures and a leak-resistant seal to protect specimens. All containers sized 20 mL through 120 mL are 95kPa compliant.