Cliniplate™ 96-Well Microplates, Thermo Scientific
Supplier: Thermo Fisher Scientific
9502227 95029330 9502887 9502867
28298-602CS 406.2 CAD
28298-602 28298-604 28298-610 28298-606
Cliniplate™ 96-Well Microplates, Thermo Scientific
Microplates Immunology Microplates and Strips
These microplates are designed for enzyme immunoassays
Ideal for use with instrumentation adapted for 96-well plates. Cliniplates™ feature excellent optical and binding precision, uniform protein binding within and between plates, and a standard raised radius well shape to avoid cross-contamination. White and black plates are designed for assays utilizing fluorescent or luminescent markers.
Ordering information: Sterile plates are individually wrapped with lids.