Immulon® Microtiter™ 96-Well Plates and Strips, Thermo Scientific
Supplier: Thermo Fisher Scientific
3355 3555 3855 6301 6404 3455 6309 6405 3655
62402-947PK 428.58 CAD
62402-947 62402-952 62402-959 62402-961 62402-963 62402-966 62402-972 62403-074 62403-076 62403-082 62402-954
Immulon® Microtiter™ 96-Well Plates and Strips, Thermo Scientific
Microplates Immunology Microplates and Strips
These plates are ideal for a variety of colorimetric techniques, such as ELISA, which are performed in a 96-well format
Immulon 1 B exhibits a consistent medium binding affinity to proteins and complexes primarily with hydrophobic tendencies. Immulon 2 HB is an irradiated plate that provides higher binding affinity. Immulon 4 HBX features a unique resin that offers maximum protein uptake with enhanced precision and guaranteed coefficient of variation (CV) of <5%.
Choose from plates, Removawell™ Assemblies and Strips for custom formatting, and Dividastrips™ Assemblies with 2 x 8 strips that separate into two 1x8 strips.
Immulon polystyrene plastics provide a consistent surface for reproducible results over a variety of binding affinities.
Plate dimensions: 12.8 x 8.6 cm (5 x 33/8")
Ordering information: For Removawell Holder that holds eight 12-well or twelve 8-well Removawell strips and Removawell Workstation, see Microtiter® Accessories for 96-Well Plates, Thermo Scientific series.