Microtiter® 96-Well Plates, Thermo Scientific
Supplier: Thermo Fisher Scientific
2205 2305-11 2605
62402-907CS 508.85 CAD
62402-907 62402-908 62402-910 62402-912 62402-914 62402-918 62402-933
Microtiter® 96-Well Plates, Thermo Scientific
Microplates Immunology Microplates and Strips
Developed for general applications, including sample storage, sample or reagent transfer, serial dilution, reagent mixing, hemagglutination, and complement fixation.
These 96-well plates are available in rigid styrene or flexible vinyl. The vinyl plates may be cut into strips or individual wells, and are recommended for incubation procedures up to 60°C (140°F).