Finnpipette™ F1 Multichannel Pipettes, Thermo Scientific
Supplier: Thermo Fisher Scientific
4661000N 4661010N 4661020N 4661030N 4661040N 4661050N 4661060N 4661070N 4661080N 4661090N 4661000N 4661010N 4661020N 4661030N 4661060N 4661080N
89080-461EA 2290.32 CAD
89080-461 89080-463 89080-465 89080-467 89080-469 89080-471 89080-473 89080-475 89080-477 89080-479 CA89080-461 CA89080-463 CA89080-465 CA89080-467 CA89080-473 CA89080-477
Finnpipette™ F1 Multichannel Pipettes, Thermo Scientific
Pipettes Micropipettes
Performance and ergonomic comfort make the Finnpipette™ F1 Multichannel Pipettes a great choice for any lab.
  • Lightweight design
  • Silent volume adjustment
  • Secure volume lock
  • Five-year warranty
Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of RSI. Enjoy the new silent and lighter volume adjustment mechanism with over 50% lower forces compared to earlier models. The secure volume setting prevents undesirable volume drift during operation.
Simply lift the plunger up to set volume and pressMore Product Information