Corning® Gel-Loading Pipette Tips, Corning
Supplier: Corning
4853 4854 4815 4884
29442-666CS 145.43 CAD
29442-666 29442-668 29443-288 29443-316
Corning® Gel-Loading Pipette Tips, Corning
Pipette Tips
These gel-loading pipette tips feature a capillary end that allows easy access into vertical and horizontal electrophoresis gels.
  • Disposable
  • Certified RNase-/DNase-free and non pyrogenic
Ordering information: Available in flat and round tip configurations. Flat tips are designed for use in 0.2 and 0.4 mm sequencing gels. Round tips are designed for use in SDS-PAGE 0.5 mm gels.
Order Now
SPECIFICATIONS
Environmentally Preferable