CELLSTAR® Cell Culture Multiwell Plates, Polystyrene, Greiner Bio-One
Supplier: Greiner Bio-One
657165 657185 662102 662165 665102 665165 677102 677165 657160 662160 665180 677180
82050-844CS 217 CAD
82050-844 82050-846 82050-888 82050-894 82050-926 82050-928 82051-000 82051-002 82050-842 82050-892 82050-930 82051-004
CELLSTAR® Cell Culture Multiwell Plates, Polystyrene, Greiner Bio-One
Microplates Cell Culture Plates
Cell Culture Multiwell Plates allow for optimal cultivation conditions for sensitive cells and cells cultivated under restricted growth conditions.
  • Free of RNase, DNase, and human DNA
  • Nonpyrogenic
  • Virgin polystyrene construction
  • Chimney well design helps reduce cross-contamination
Surface treatments provide the conditions for improved cell adherence, accelerated cell expansion, and they increase overall cell yield.
Manufactured from optically clear virgin polystyrene, these flat-bottom plates are supplied sterile and pyrogen-free with a printed lot number for traceability. The chimney well design and low evaporation lids with condensation rings reduce the risk of cross-contamination. Plates are available tissue culture treated for the culture of anchorage-dependent cells or nontreated for suspension cell culture applications. Excellent heat exchange within each plate is ensured during incubation through air circulation spaces between stacked plates. Uniform design makes them suitable for manual and automated handling. The well depth and diameter also allows the use of specific tissue culture inserts.
Sterilized by radiation. DNase-, RNase-, and human DNA-free. Nonpyrogenic and noncytotoxic.
Note: The nontreated, 24-well plates packaged 12 per sleeve are black, nonsterile, and supplied without lid.
Ordering information: Supplied in sterile and user friendly packaging.