Malachite green solution
Supplier: G-Biosciences
786-2120
77540-298EA
48.74
CAD
77540-298
Malachite green solution
Malachite green solutions
Malachite Green is used as a counterstain.
- Quick staining dye
It differentiates acid-fast bacteria from non-acid-fast bacteria