Carbol fuchsin solution (Ziehl-Neelsen)

Supplier: G-Biosciences

786-2125 786-2126
77540-308EA 48.74 CAD
77540-308 77540-310
Carbol fuchsin solution (Ziehl-Neelsen)
Carbol fuchsin

Ziehl-Neelsen Carbolfuchsin is used to detect Cryptosporidium spp.


  • Quick staining dye


It differentiates acid-fast bacteria from non-acid-fast bacteria

Formula: C₂₁H₂₂ClN₃
MDL Number: MFCD00013129
CAS Number: 4197-24-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR