Gram Stain Enhancer
Supplier: G-Biosciences
786-2090
786-2091
77540-238EA
316
CAD
77540-238
77540-240
Gram Stain Enhancer
Stains
Microscopy and staining reagents
Gram Enhancer is used to reduce the background noise during staining process.
- An aqueous solution of Fast Green, Tartrazine and Formaldehyde
Quick constrasting of Gram-negative bacteria