Giemsa's stain kit

Supplier: G-Biosciences

Synonyms: Giemsa's azur-eosin-methylene blueAzure mixture sicc. Giemsa stainAzur-Eosin-Methylene blue acc. Giemsa

786-2078
77540-214KT 237.6 CAD
77540-214
Giemsa's stain kit
Giemsa's stain

Giemsa Stain Kit is a modified Wright stain.


Use for detection and indentification of parasites from blood and tissues


Packaging: Kit includes: Giemsa Fixative, Giemsa Reagent A and Giemsa Reagent B

MDL Number: MFCD00081642
CAS Number: 51811-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR