Kinyoun AFB Stain Kit
Supplier: G-Biosciences
786-2117
77540-292EA
298.7
CAD
77540-292
Kinyoun AFB Stain Kit
Kinyoun AFB Stain Kit
Kinyoun AFB Stain kit is used to detect Cryptosporidium spp.
It differentiates acid-fast bacteria from non-acid-fast bacteria
Packaging: Kit includes two bottles of 250 ml each Kinyoun Carbolfuchsin, Decolorizer and Methylene Blue.