Starter™ 2200 pH Bench Meter
Supplier: OHAUS CORP.
30655944 30656033 30655944
470336-580EA 736.2 CAD
470336-580 77679-478 77682-010
Starter™ 2200 pH Bench Meter
Electrochemistry Meters pH and Multi-Parameter Meters
Designed with a clear, user-friendly interface and the ability to store up to 99 items in its internal memory, the Starter™ 2200 pH bench meter is a reliable and efficient addition to basic laboratories or science classrooms.
  • Affordable, intuitive pH measurement instrument
  • 3.5 inch segment LCD with backlight
  • AC adapter (included)
  • ABS housing, mechanical keys, built-in electrode holder
  • Auto buffer recognition, two point calibration, electrode condition icon
Starter™ 2200 pH bench meter also features auto-buffer recognition and auto-endpoint modes that make measurement and calibration simple, straightforward and efficient.