Corning® BioCoat® Gelatin Culture Dishes, Corning

Supplier: Corning
356653 354653
CA62405-920CS 462.91 CAD
CA62405-920 CA62405-922
Corning® BioCoat® Gelatin Culture Dishes, Corning
Culture Dishes Cell Culture Dishes
Corning BioCoat Gelatin cultureware provides an attachment and growth promoting substrate for the culture of a variety of cell types. Gelatin is commonly used in the culture of vascular endothelial cells, muscle, embryonic stem (ES) cells, and F9 teratocarcinoma cells. It is also suitable for promoting adhesion of transfected cell types. Gelatin is a heterogeneous mixture of water-soluble proteins derived through the hydrolysis of Collagen.

  • Promotion of cell attachment and spreading
  • Culture of normal and transfected F9 teratocarcinoma cells for gene expression studies
  • Culture of HUVEC for E-Selectin7 expression and VEGF induction8
  • Source: Gelatin, porcine
  • Storage: 4 to 30 °C under dry conditions
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR