Neptune® BT Barrier Pipette Tips, Pre-Sterile
Supplier: Biotix
63300746 63300748 63300751 63300753 63300752 63300757 63300759 63300754 63300763 63300755 63300756 63300749 63300750 63300764 63300762
89140-160CS 1164.93 CAD
89140-160 89140-162 89140-164 89140-166 89140-930 89140-932 89140-936 89168-750 89168-752 89168-754 89168-756 89217-468 89217-470 10144-786 10144-826
Neptune® BT Barrier Pipette Tips, Pre-Sterile
Pipette Tips
Pre-sterile universal barrier tips are made from virgin polypropylene and feature high density polyethylene filters to ensure against aerosol contamination and provide superior consistency.
  • Trays are autoclavable
  • Enabling reuse for maximum sustainability
Eco-friendly empty pipette tip trays are manufactured from recycled polypropylene. When combined with ESP™ reload systems, these tip trays reduce plastic waste by up to 90%. The trays provide superior stability, firm placement, and a maximum space saving design.
Thin wall design provides an optimal seal for single and multichannel pipettors. Tips featuring the S³ polymer eliminate sample hold up and ensure a more precise sample draw and dispense. Eco-friendly racks are made from recycled polypropylene and feature a unique compact packaging configuration.
Certifications: Certified free of RNase, DNase, pyrogens, and PCR Inhibitors. Manufactured under stringent controls in an ISO 9001 certified facility. Each and every lot is tested for accuracy and precision, ensuring superior quality and compatibility. All raw materials are controlled.