Corning® BioCoat™ Matrigel® Matrix Cellware, Corning
Supplier: Corning
354432 354433 354603 354605 354607
CA-CB354432CS 720.66 CAD
CA-CB354432 CA-CB354433 CA-CB354603 CA-CB354605 CA-CB354607
Corning® BioCoat™ Matrigel® Matrix Cellware, Corning
Microplates Cell Culture Plates
Corning® BioCoat™ Matrigel® Matrix Cellware consists of Falcon® Multiwell Tissue Culture inserts, plates, and dishes treated with a uniform application of Corning® Matrigel® Basement Membrane Matrix to promote differentiation of a variety of cell types
Corning® Matrigel® source: Engelbreth-Holm-Swarm (EHS) mouse tumor. Tested for ability to promote neurite outgrowth from chick dorsal root ganglia (in the absence of NGF). Tested and found negative for bacteria and fungi.
Cellware is stable for >=3 months from date of shipment when stored at –20°C (–4°F); keep frozen until use. Thin-layer cellware is stable for >=3 months when stored at 2 to 8°C (35.6 to 46.4°F). Inserts stable for >=3 months at –20°C (–4°F); keep frozen until use.
Convenient, ready-to-use cellware offers lot-to-lot consistency and reliable cell culture performance. Use Corning® BioCoat™ Matrigel® Matrix Cellware for the culture of muscle cells, epithelial cells, endothelial cells, and more.
Ordering information: Inserts are packaged six per 6-well plate and twelve per 24-well plate.