Corning® IsoTip™ Pipette Tips, Corning
Supplier: Corning
4801 4823 4807 4808 4809 4821 4810
10010-364CS 293.24 CAD
10010-364 29442-620 29443-272 29443-274 29443-276 29442-616 29443-278
Corning® IsoTip™ Pipette Tips, Corning
Pipette Tips
These pipette tips feature an inert, hydrophobic bonded-fiber barrier that prevents aerosolised contaminants from coming in contact with air displacement pipettors. Ideal for applications where avoidance of cross contamination is critical, such as DNA amplification and radioisotope handling.
  • Exceptional durability
  • All Corning® tip racks stand up to the demands of use with multichannel pipettors and withstand repeated autoclaving cycles
  • Sterile and certified RNase-/ DNase-free and non pyrogenic
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification I-RECs, ISO 14001 - Reynosa